Learn More
Medchemexpress LLC Nourseothricin sulfate | 96736-11-7 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12906510MM/1ML
Encompass_Preferred
Nourseothricin sulfate is a broad-spectrum antibiotic that works by destroying the outer membrane of Gram-negative bacteria. It also functions as a dominant selective marker for Fonsecaea pedrosoi. This antibiotic inhibits protein biosynthesis in prokaryotic cells and strongly suppresses the growth of eukaryotic organisms such as fungi.
  • Broad-spectrum antibiotic
  • Destroys the outer membrane of Gram-negative bacteria
  • Functions as a dominant selective marker for Fonsecaea pedrosoi
  • Inhibits protein biosynthesis in prokaryotic cells
  • Suppresses the growth of eukaryotic organisms such as fungi
  • Can be used as a selective marker for bacteria, yeast, filamentous fungi, and plant cells
Catalog No. 50-003-38931