Learn More
Cayman Chemical NourseothrIcInsulfate 10mg
SDP
Supplier:  Cayman Chemical 1622710 MG
Encompass_Preferred
A broad-spectrum antibiotic that inhibits protein synthesis by inducing miscoding; commonly used as a dominant selection antibiotic for genetically modified bacteria, yeasts, fungi, protozoa, plants, and mammalian cells
Catalog No. 50-297-0310