Learn More
Electron Microscopy Sciences ProPlate™ Oncyte Nova 384 NC Square Pad, 2.5x2.5mm Glass Substrate
SDP
Supplier:  Electron Microscopy Sciences 6348527
Encompass_Preferred
Patented ProPlate technology for high throughput robotic processing of microarrays. Grace BioLabs developed bottomless SBS compliant microtiter plate superstructures for use with robotic processors. Features guaranteed flat SBS compliant microtiter plate format which enables robotic processing of assays. It is compatible with glass or plastic substrates and has a water-tight seal between superstructure and substrate. With ONCYTE Nitrocellulose Coated Substrates, 2.5 x 2.5 mm for SBS Microtiter Plate, Glass Substrate.
Catalog No. 50-343-37