Learn More
Waters Corp Nova-Pak C18 Radial-Pak Column, 60Å, 4 µm, 8 mm X 100 mm, 1/pk
Supplier:  Waters Corp WAT086342
Encompass_Preferred
Nova-Pak C18 columns are general purpose, silica-based, reversed-phase C18 columns that are based on 4 µm particle technology. Compared to a 5 µm column, Nova-Pak columns offer higher resolution, more efficient separations, and faster chromatography.
Catalog No. 50-818-677