Learn More
Waters Corp HPLC Column, 4 μm, 5 x 100 mm, C18
Supplier:  Waters Corp WAT080100
Encompass_Preferred
The 4 μm small particle size of Nova-Pak bonded phases offers high resolution, more efficient and faster chromatography by using shorter columns, reducing solvent consumption or by using longer columns to resolve even the most complex mixture.
  • The 4 μm small particle size offers high resolution, more efficient and faster chromatography.
  • Can be used with shorter columns, reducing solvent consumption.
  • Can be used with longer columns to resolve complex mixtures.
  • Semi-preparative columns are available with 6 μm particle size packings for a range of separation possibilities.
  • Stringent QC procedures in a cGMP manufacturing facility ensure batch-to-batch reproducibility.
Catalog No. 50-785-899