Learn More
Waters Corp Waters Column, C18, 4 μm, 3.9 mm, 300 mm
Supplier:  Waters Corp WAT011695
Encompass_Preferred
These general-purpose, silica-based, reversed-phase C18 columns utilize 4 μm particle technology for high-performance liquid chromatography (HPLC) systems. They deliver higher resolution, more efficient, and faster separations compared to 5 μm columns. The smaller particles reduce solvent consumption while maintaining the ability to resolve complex mixtures.
  • Achieve high resolution and fast, efficient chromatography.
  • Reduce solvent consumption by using shorter column lengths with smaller particles.
  • Benefit from a larger surface area for enhanced interaction and separation of elutes.
  • Experience faster separations, producing concentrated fractions and reducing operating costs.
  • Ensure high reproducibility between batches and columns through strict quality control.
  • Optimize column length for desired results, allowing for higher flow rates or higher resolution.
Catalog No. 50-785-046