Learn More
Waters Corp WATERS CORPORATION HPLC Column, C18, 4 μm, 4.6 mm X 150 mm
Supplier:  Waters Corp WAT044375
Encompass_Preferred
These C18 columns are silica-based, reversed-phase columns featuring 4 μm particle technology. They offer higher resolution, more efficient separations, and faster chromatography than 5 μm columns, suitable for various system configurations and sample types.
  • Higher resolution, more efficient, and faster separations compared to 5 μm columns.
  • Reduced solvent consumption due to smaller particles.
  • More interaction time for enhanced separation.
  • Innovative 4μ particle technology provides possibilities for high efficiency and carbon surface area.
  • Operates across a pH range of 2 to 8.
  • End-capped for optimal security and ease of use.
  • Manufactured under strict quality control processes for high batch-to-batch reproducibility.
Catalog No. 50-785-571