Learn More
Waters Corp Nova-Pak Column, Cyano (CN), 60Å, 4 μm, 3.9 mm x 150 mm
Supplier:  Waters Corp WAT044245
Encompass_Preferred
Nova-Pak HP CN columns are silica-based, cyano phases featuring 4 μm particle technology. These columns offer unique compound selectivity for both normal-phase and reversed-phase applications. Their end-capped design prevents sample loss, maximizing efficiency and extending lifetime.
  • Silica-based, cyano phases with 4 μm particle technology.
  • Unique compound selectivity for normal-phase and reversed-phase applications.
  • Excellent separation of polar compounds.
Catalog No. 50-785-565