Learn More
Waters Corp HPLC Cartridge, Silica, 60Å, 4 μm, 5x100 mm
Supplier:  Waters Corp WAT010986
Encompass_Preferred
This HPLC Cartridge Radial-Pak Nova-Pak Silica is designed for high-performance liquid chromatography applications. It features a robust silica material with a specific pore size and particle size, ensuring efficient and reliable separations.
  • Particle size: 4 μm
  • Pore size: 60Å
  • Dimensions: 5x100 mm
  • Material: Silica
Catalog No. 50-784-996