Learn More
Cytiva NOVASIP EMFLON PFR 0.2UM DRAIN
Supplier:  Cytiva C3PFRP1
Encompass_Preferred
Emflon PFR membrane in Novasip capsule 0 2 m sterilizing grade 2300 cm EFA quick connect and disconnect coupling vent 4-6 mm hose barb drain with valve
Catalog No. 50-003-30570