Learn More
Alkali Scientific Novobiocin Sodium Salt, 1 G
SDP
Supplier:  Alkali Scientific NOV002CP
Encompass
Novobiocin Sodium Salt is a broad-spectrum antibiotic available in a 1g powder form. It is used primarily for bacterial inhibition in laboratory settings, particularly for research on Gram-positive bacteria. Novobiocin works by inhibiting bacterial DNA gyrase, preventing DNA replication and transcription. It is commonly used in microbiological studies to assess bacterial resistance mechanisms, investigate DNA replication processes, and test for the efficacy of combination antibiotic therapies. This compound is also used in clinical research for the development of treatments for infections caused by antibiotic-resistant bacteria. The 1g quantity is suitable for preparing concentrated solutions for laboratory experiments.
Catalog No. 50-006-60897