Learn More
Medchemexpress LLC Novobiocin (Sodium) | 1476-53-5 | 99.1% | 1 ML
Supplier: Medchemexpress LLC HYB0425A10MM/1ML
Novobiocin sodium is a potent and orally active antibiotic that functions as a DNA gyrase inhibitor and a heat shock protein 90 (Hsp90) antagonist. It shows potential for research into highly beta-lactam-resistant pneumococcal infections and demonstrates anti-orthopoxvirus activity. This product is intended for research use only.
- Potent and orally active antibiotic
- Acts as a DNA gyrase inhibitor
- Functions as a heat shock protein 90 (Hsp90) antagonist
- Useful in research for highly beta-lactam-resistant pneumococcal infections
- Exhibits anti-orthopoxvirus activity
- Inhibits ATP binding to gyrase B
- Induces caspase-3/7 enzyme-dependent apoptosis in K562, HL60, and Mutz-2 cells