Learn More
Cayman Chemical NovobIocInsodIum salt 5g
SDP
Supplier:  Cayman Chemical 184575 G
Encompass_Preferred
An inhibitor of DNA gyrase; inhibits the ATPase activity of DNA gyrase, permitting relaxation of negative supercoils; has been used to generate positively supercoiled DNA; inhibits the chaperone activity of Hsp90 (IC50 = 700 mM)
Catalog No. 50-296-7678