Learn More
Selleck Chemical LLC NOX-6-18 E4773-1G
SDP
Supplier:  Selleck Chemical LLC E47731G
Encompass_Preferred
NOX-6-18 is a potent and selective antagonist of GPR132 with an IC50 of 17 nM It regulates macrophage reprogramming within pancreatic islets reduces weight gain and improves glucose metabolism in mice on a high-fat diet
Catalog No. 50-000-39545