Learn More
Biotang Inc NP-40, 100 ML, P-877, NP-40 (Nonidet P-40 Substitute), CAS # 9036-19-5. Viscous liquid. Soluble in H2O
Supplier:  Biotang Inc BTBB914
Encompass_Preferred
NP-40, 100 ML, P-877, NP-40 (Nonidet P-40 Substitute), CAS # 9036-19-5. Viscous liquid. Soluble in H2O. Nonionic non-denaturing detergent is variously used in molecular biological, cell biological, biochemical studies..
Catalog No. 50-152-368