Learn More
Cayman Chemical NPalmitoyl Glycine
SDP
Supplier:  Cayman Chemical 1000902050 MG
Encompass_Preferred
an 18-carbon saturated fatty acid that is structurally similar to the phospholipid-derived N-acyl ethanolamines; inhibits heat-induced firing of nociceptive neurons in rat dorsal horn; induces transient calcium influx in native dorsal root ganglion (DRG) cells and in the PTX-sensitive, DRG-like cell line F-11 (EC50 = 5.5 µM)
Catalog No. 50-288-8913