Learn More
Enzo Life Sciences NPC-15437 . dihydrochloride (1mg). CAS: 136449-85-9; 141774-20-1
SDP
Supplier:  Enzo Life Sciences ALX270104M001
Encompass_Preferred
PKC inhibitor. Selective inhibitor of protein kinase C (PKC). Alternative name: (S)-2,6-diamino-N-{[1'-(1''-oxotridecyl)-2'-piperidinyl]methyl}hexanamide . 2HCl. Formula: C25H50N4O2 . 2 HCl. MW: 511.6. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water, DMSO or methanol. Long Term Storage: +4°C. Handling: Hygroscopic.
Catalog No. 50-200-8406