Learn More
Cayman Chemical NPC-15437hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 202125 MG
Encompass_Preferred
A selective PKC inhibitor (IC50 = 19 µM); inhibits phorbol ester- (Ki = 5 µM) and phosphatidylserine-induced (Ki = 12 µM) PKC activity but does not affect the activity of cAMP-dependent or Ca2+/calmodulin-dependent protein kinases
Catalog No. 50-298-3292