Learn More
Enzo Life Sciences Nuclear pore complex proteins monoclonal antibody (MAb414) (purified) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1050100
Encompass_Preferred
Clone: MAb414. Isotype: IgG1. Immunogen: Nuclear pore complex mixture. Species reactivity: Yeast, Xenopus, Vertebrate. Specificity: Believed to recognize the repeated FXFG repeat sequence in nuceloporins including p62, p152 and p90 and other proteins. Applications: ICC, IHC, IP, WB Immunoelectron microscopy. Purity: Protein G purified. Formulation: Liquid. In PBS containing 0.03% thimerosal.
Catalog No. 50-201-2665