Learn More
Medchemexpress LLC Npd10084 PKM2 inhibitor | 1040706-91-9 | 99.7% | 345.39 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14861510MM/1ML
Encompass_Preferred
NPD10084 is a pyruvate kinase PKM2 inhibitor that inhibits non-glycolytic signaling in cancer cells. It disrupts the interaction between PKM2 and β-catenin or STAT3, thereby inhibiting downstream signaling.
  • Demonstrates antiproliferative activity against colorectal cancer cells both in vitro and in vivo
  • Targets pyruvate kinase muscle isoform 2 (PKM2)
Catalog No. 50-003-60451