Learn More
Selleck Chemical LLC Plinabulin (NPI-2358) 50mg 714272-27-2
SDP
Supplier:  Selleck Chemical LLC S117650MG
Encompass_Preferred
Plinabulin (NPI-2358) is a vascular disrupting agent (VDA) against tubulin-depolymerizing with IC50 of 9.8~18 nM in tumor cells. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-887-3