Learn More
Enzo Life Sciences NPPB (10mg). CAS: 107254-86-4
SDP
Supplier:  Enzo Life Sciences BMLCM1000010
Encompass_Preferred
A potent chloride channel blocker (IC50=0.1-100 µM depending on channel subtype and assay method). Inhibits cyclooxygenase (COX; IC50=8 µM). Evidence of mitochondrial uncoupling in mouse peritoneal macrophages, due to protonophore activity. Alternative name: 5-Nitro-2-(3'-phenylpropyl-amino)benzoic acid. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml; warm). Long Term Storage: Ambient.
Catalog No. 50-201-0411