Learn More
Medchemexpress LLC NPPB Human His 50ug
SDP
Supplier:  Medchemexpress LLC HYP71119A50UG
Encompass_Preferred
Recombinant human NPPB (B-type natriuretic peptide) with an N-terminal 6xHis tag, expressed in E. coli and supplied as a filtered solution. The construct corresponds to residues H27-H134 (≈16.0 kDa) and is provided in a stabilizing buffer at an example concentration of ~1.28 mg/mL. This reagent is intended for cardiovascular biology research, peptide signaling studies, and assay development that require purified, tagged human NPPB.
  • Recombinant human sequence H27-H134 representing the mature peptide region.
  • N-terminal 6xHis tag compatible with affinity purification and detection.
  • Expressed in E. coli for consistent, high-yield production.
  • Supplied as a 0.22 μm filtered solution for convenience and reduced contamination risk.
  • Formulated in pH 7.4 buffer with 20% glycerol to enhance stability during storage.
  • Provided at a concentration suitable for downstream biochemical and cell-based assays.
Catalog No. 50-001-96567