Learn More
Medchemexpress LLC NPPB Human His so 10ug
SDP
Supplier:  Medchemexpress LLC HYP7111910UG
Encompass_Preferred
Recombinant human NPPB (B-type natriuretic peptide) is supplied as an N-terminal 6xHis-tagged solution expressed in E. coli and formulated for stability and immediate use in cardiovascular and signaling research. The preparation is highly pure, low in endotoxin, and provided at a defined concentration in a buffered glycerol-containing formulation for reliable handling and storage.
  • High purity (>98%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
  • Ready-to-use, 0.22 μm filtered solution in physiological buffer.
  • N-terminal His tag enables detection or affinity capture.
  • Stable frozen formulation; aliquot and avoid repeated freeze-thaw cycles.
Catalog No. 50-001-96552