Learn More
Medchemexpress LLC NPR-C activator 1 | 2768328-61-4 | 99.3% | C18H24N6O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14331610MG
Encompass_Preferred
NPR-C activator 1 is a small-molecule agonist of the natriuretic peptide receptor C (NPR-C) developed for research use. It is reported to have an EC50 of ≈ 1 μM, a molecular formula of C18H24N6O3, and high analytical purity. The compound is provided as a solid and is soluble in DMSO for preparation of concentrated stock solutions.
  • Potent NPR-C agonist (EC50 ≈ 1 μM).
  • High purity (99.3%).
  • Soluble in DMSO at 100 mg/mL for stock preparation.
  • Supports in vivo formulation at 2.5 mg/mL per supplier protocols.
  • Suitable as a research tool for NPR-C signaling studies.
Catalog No. 50-002-61023