Learn More
Cayman Chemical nprIlysInCD10N-Term Rabb 100uL
SDP
Supplier:  Cayman Chemical 32276100 UL
Encompass_Preferred
An adenosine A2A and A2B receptor dual antagonist inhibits the ability of adenosine to suppress activation of human CD4 or CD8 T cells as well as monocyte-derived dendritic cells in vitro reduces tumor growth in an AT3-OVA syngeneic mouse model alone or in combination with doxorubicin acts synergistically with an anti-PD-1 antibody to reduce tumor growth in a B16/F10 syngeneic mouse model
Catalog No. 50-294-8530