Learn More
Medchemexpress LLC NPS-1034 | 1221713-92-3 | 98.0% | 551.54 g/mol | C31H23F2N5O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10050925MG
Encompass_Preferred
NPS-1034 is a small-molecule dual inhibitor of the receptor tyrosine kinases AXL and MET, used in research to probe MET/AXL-driven signaling and resistance mechanisms. It exhibits low-nanomolar potency against both targets and is provided as a high-purity powder for in vitro and preclinical studies.
  • Dual inhibition of AXL and MET for pathway studies.
  • Low-nanomolar potency: AXL IC50 = 10.3 nM, MET IC50 = 48 nM.
  • High purity supports reproducible experimental results.
  • Supplied as a powder with defined storage conditions.
  • Suitable for in vitro kinase assays and cell-based studies.
  • Available in small research quantities for preclinical use.
Catalog No. 50-002-14737