Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NPS-2143 hydrochloride | 324523-20-8 | MFCD16038896 | 100.0% | 445.38 g·mol⁻¹ | C24H26Cl2N2O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY1017110MG

Encompass_Preferred

NPS-2143 hydrochloride is an orally active calcilytic agent and a selective, potent antagonist of the calcium-sensing receptor (CaSR). It inhibits CaSR-mediated increases in cytoplasmic Ca2+ (reported IC50 ≈ 43 nM) and is used as a research reagent for studies of CaSR signaling, pharmacology, and related cellular assays.

  • Selective CaSR antagonist with reported IC50 ≈ 43 nM.
  • High purity (≈99.98%).
  • Solid, white to off-white powder suitable for laboratory use.
  • Molecular weight 445.38 g·mol⁻¹; molecular formula C24H26Cl2N2O2.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Available in small research pack sizes (for example, 10 mg).

Catalog No. 50-002-46997


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.