Learn More
Abcam NPS 2143 hydrochloride, Ca2+-sensing receptor antagonist, 50MG
Supplier:  Abcam AB14505050MG
Encompass_Preferred
MW 445.4 Da, Purity >99%. Potent, selective Ca2+-sensing receptor antagonist (IC50 = 43 nM). Stimulates parathyroid hormone secretion (EC50 = 41 nM). Shows antihypertensive effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5446