Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NPS 2390 | 226878-01-9 | 99.9% | 307.39 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY1109510MG

Encompass_Preferred

NPS 2390 is a noncompetitive antagonist of mGluR1 and mGluR5. It is also identified as a potent inhibitor of CaSR (calcium-sensing receptor). This product is intended for research use only and is not sold to patients.

  • Noncompetitive antagonist of mGluR1 and mGluR5.
  • Potent CaSR (calcium-sensing receptor) inhibitor.
  • Available in various quantities (5 mg, 10 mg, 25 mg, 50 mg, 100 mg solid; 10 mM * 1 mL in DMSO solution).
  • High purity: 99.90%.

Catalog No. 50-003-44026


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.