Learn More
ABclonal Technology NQO2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A11746500UL
Encompass_Preferred
This gene encodes a member of the thioredoxin family of enzymes. It is a cytosolic and ubiquitously expressed flavoprotein that catalyzes the two-electron reduction of quinone substrates and uses dihydronicotinamide riboside as a reducing coenzyme. Mutations in this gene have been associated with neurodegenerative diseases and several cancers. Alternative splicing results in multiple transcript variants.
Catalog No. 50-000-99149