Learn More
Medchemexpress LLC NR2F6 modulator-1 | 904449-84-9 | >99.46% | 419.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1472285MG
Encompass_Preferred
NR2F6 modulator-1 is a potent nuclear receptor subfamily 2, group F, member 6 (NR2F6) modulator. This compound is valuable for research into immune modulation and the activity of cancer stem cells.
  • Potent nuclear receptor subfamily 2, group F, member 6 (NR2F6) modulator
  • Useful for immune modulation research
  • Assists in modulation of cancer stem cell activity research
Catalog No. 50-003-61973