Learn More
Cayman Chemical NR6A1humnrecombInt 100ug
SDP
Supplier:  Cayman Chemical 41073100 UG
Encompass_Preferred
Source: Recombinant human NR6A1 expressed in E. coli • Amino acids: 2-480 (full length) • MW: 54.42 kDa
Catalog No. 50-294-5950