Learn More
Medchemexpress LLC Nrf2 activator-2 | 2770448-53-6 | 99.2% | C20H17BrO3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY145879
Encompass_Preferred
Nrf2 activator-2 (compound O15), an Osthole derivative, is a potent Nrf2 agonist with an EC50 of 2.9 μM in 293 T cells. This compound effectively inhibits the interaction between Keap1 and Nrf2, thereby activating Nrf2 and leading to a marked decrease in the level of ubiquitinated Nrf2 in cells.
  • Potent Nrf2 agonist.
  • Effectively inhibits Keap1-Nrf2 interaction.
  • Activates Nrf2 signaling pathway.
  • Decreases ubiquitinated Nrf2 levels.
Catalog No. 50-003-57190