Learn More
Medchemexpress LLC NRG1-beta 2 Human 2ug
SDP
Supplier:  Medchemexpress LLC HYP733042UG
Encompass_Preferred
HRG1- 1 belongs to a family of structurally-related polypeptide growth factors derived from alternatively spliced genes induces Fn14 expression in MCF7 cells NRG1-beta 2 Protein Human is the recombinant human-derived NRG1-beta 2 protein expressed by E coli with tag free
Catalog No. 50-001-79243