Learn More
Aviva Systems Biology Corp NRIP2 Antibody - N-terminal region (ARP50702_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50702_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRIP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NRIP2. Alternate names: DKFZP761G1913ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60414