Learn More
AllCells LLC Peripheral Blood- Cryopreserved- CD19+ B Cells- Negatively Selected- 10M
Supplier:  AllCells LLC LP CR CD19 NS 10M
Encompass
CD19+ B cells are negatively selected from peripheral blood leukapheresis using immunomagnetic microbeads targeting non-B cells. CD19+ B cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 85%. Viability is ≥ 80% post-thaw.
Catalog No. NC2742222