Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Norprene Food-Grade, 1/4"" ID x 3/8"" OD; 50 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9571030
Encompass
This tubing is ideal for the food, dairy, and beverage industries. Characteristics include heat, ozone, and UV light resistant; non-aging; nonoxidizing; superior acid and alkali resistance. Tubing is manufactured according to GMP and has a temperature range of ¿60 to 275¿F (¿51 to 135¿C). Sterilize with autoclave.
Catalog No. NC9075794