Learn More
Medchemexpress LLC NRX-0492 | 2416130-57-7 | C43H51N11O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY153357100MG
Encompass_Preferred
NRX-0492 is an orally active PROTAC-class BTK degrader that catalyzes BTK ubiquitination and proteasome degradation. It inhibits B cell receptor (BCR) mediated signaling, transcription programs, and chemokine secretion, demonstrating antitumor effects against chronic lymphocytic leukemia.
  • Catalyzes BTK ubiquitination and proteasome degradation
  • Inhibits B cell receptor (BCR) mediated signaling
  • Inhibits transcription programs and chemokine secretion
  • Demonstrates antitumor effects against chronic lymphocytic leukemia
  • Shows high-affinity binding to wild-type and C481S/T474I mutant BTK
  • Induces BTK degradation in CLL cells from blood and spleen in vivo
  • Reduces proportion of Ki67+/CD69+ cells in vivo
  • Decreases splenic tumor burden in CLL patient-derived xenografts
Catalog No. 50-003-73410