Learn More
TARGETMOL CHEMICALS INC NRX-252114 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T635165MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. NRX-252114 (NRX252114) induces degradation of mutant (beta)-catenin NRX-252114 is a potent enhancer of the interaction of (beta)-catenin with the homologous E3 ligase SCF(beta)-TrCP and is able to enhance the binding of pSer33/S37A (beta)-catenin to (beta)-TrCP with an EC50 of 6.5 nM and a Kd of 0.4 nM. Kd is 0.4 nM. purity: 98%
Catalog No. 50-257-9420