Learn More
Medchemexpress LLC Nrx-2663 | 2763260-34-8 | 98.0% | C20H13F3N2O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14144810MG
Encompass_Preferred
NRX-2663 is a small-molecule research compound that enhances the interaction between β-catenin and its E3 ligase SCFβ-TrCP, and is used in biochemical and cell-based assays to probe β-catenin binding and degradation. Reported activity includes an EC50 of 22.9 μM and a Kd of 54.8 nM.
  • Enhances β-catenin and SCFβ-TrCP interaction.
  • Reported EC50 22.9 μM and Kd 54.8 nM.
  • High purity: 98.0%.
  • Suitable for biochemical and cell-based assays.
  • Available in multiple small-scale packaging options, including a DMSO solution format.
Catalog No. 50-002-58762