Learn More
Medchemexpress LLC NRX-2663 | 2763260-34-8 | 99.0% | C20H13F3N2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141448100MG
Encompass_Preferred
NRX-2663 is an enhancer of the interaction between β-catenin and its cognate E3 ligase, SCFβ-TrCP. It enhances the binding of β-catenin peptide for β-TrCP with an EC50 of 22.9 μM and a Kd of 54.8 nM. It also significantly enhances the ubiquitylation of the pSer33/Ser37 β-catenin peptide, making it suitable for laboratory research.
  • Enhances binding of β-catenin peptide for β-TrCP with an EC50 of 22.9 μM
  • Shows improved potency in binding assay with pSer33/Ser37 β-catenin (EC50 of 80 μM)
  • Provides 13-fold cooperativity of binding
  • Significantly enhances ubiquitylation of the pSer33/Ser37 β-catenin peptide
Catalog No. 50-003-53786