Learn More
Medchemexpress LLC NS-102 50mg | 136623-01-3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11442750MG
Encompass_Preferred
NS-102 is a selective kainate (GluK2) receptor antagonist and a potent GluR6/7 receptor antagonist supplied as a purified research chemical for neuroscience studies. The product is provided with analytical documentation and safety information to support laboratory use.
  • Selective kainate (GluK2) receptor antagonist.
  • Potent GluR6/7 receptor antagonist.
  • Purity approximately 98% as reported by the supplier.
  • Off-white to yellow solid available in multiple small pack sizes for research use.
  • Documentation includes certificate of analysis and safety data sheet.
Catalog No. 50-002-27407