Learn More
TARGETMOL CHEMICALS INC NS 1738 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T788425MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NS 1738 (NSC-213859) is a positive allosteric modulator of the alpha7-containing neuronal nicotinic acetylcholine receptors (nAChRs) Purity 98%
Catalog No. 50-287-6781