Learn More
Medchemexpress LLC NS-638 | 150493-34-8 | MFCD00920837 | 99.8% | 325.72 | C15H11ClF3N3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101428100MG
Encompass_Preferred
NS-638 is a small nonpeptide Ca2+-channel blocker used in research to study voltage-gated calcium channel-mediated signaling. It inhibits K+-stimulated intracellular calcium elevation (reported IC50 ≈ 3.4 μM) and shows activity in synaptosome uptake and neuronal release assays. Provided as a solid with high reported purity, it is intended for laboratory research use only.
  • Blocks K+-stimulated intracellular Ca2+ elevation (IC50 ≈ 3.4 μM).
  • Shows activity in synaptosome uptake and AMPA-stimulated GABA release assays.
  • Solid form, high purity suitable for biochemical assays.
  • Molecular weight 325.72, formula C15H11ClF3N3.
  • For research use only; not for human or veterinary applications.
Catalog No. 50-002-47133