Learn More
Mitegen Cryo-EM Autogrid Inspection Tool by Nanosoft, Low-Cost Pre-Loading Verification, Ensures Secure Grid Clipping for Autoloaders
SDP
Supplier:  Mitegen MCEMNSAGIT01
Encompass
This Cryo-EM Autogrid Inspection Tool by Nanosoft offers a crucial, low-cost verification step before loading autogrids into cassettes, vital for high-throughput cryo-electron microscopy workflows. It effectively prevents potential issues arising from improperly clipped autogrids, which can otherwise lead to significant instrument downtime. The tool is designed to fit securely into a grid button spot within a cassette loading station, remaining stable due to its spring-loaded detent. Users can easily place autogrids into the tool's groove and quickly flip them to confirm secure clipping. This final check helps ensure only well-prepared grids proceed into the autoloader system. Crafted from durable anodized aluminum, this inspection tool supports a seamless cryo-EM workflow, promoting efficiency and reliable imaging results by safeguarding instrument operation from grid-related disruptions.
Catalog No. NC1816916