Learn More
Mitegen Cryo-EM Tweezers for Vitrobot From Nanosoft, Spring-Loaded Plunge-Clamp, Modified Reverse Action, Effortless Grid Handling & Plunge-Freezing
SDP
Supplier:  Mitegen MCEMNSCT01
Encompass
Cryo-EM Tweezers by Nanosoft revolutionize the cryo-TEM sample preparation process, offering dependable and effortless operation with all generations of the Vitrobot. These fully assembled tweezers incorporate a spring-loaded Plunge-Clamp, enabling easy attachment and removal from the plunger. The clamp self-centers, ensuring precise alignment of the tweezers axis with the plunger rod for safe, controlled plunging into liquid ethane, effectively preventing common sample grid crashes. Additionally, the modified reverse action tweezers securely grasp delicate TEM grids in their natural state, requiring only a small pinch to release them into the grid box. This innovative mechanism eliminates the need for cumbersome O-rings, providing a smoother, less stressful experience for all users. Their refined design allows easy fitting between blotting pads without interference, leading to fewer lost grids and more successful imaging, significantly streamlining your cryo-EM workflow.
Catalog No. NC1816919