Learn More
Fine Science Tools Scalpel Blades - #20, Curved, 100/pk
SDP
Supplier:  Fine Science Tools 1002000
Encompass
Product: Scalpel Blades - #20/ Blade Shape: Curved/ Cutting Edge: 30mm/ Thickness: 0.4mm/ Alloy / Material: Carbon Steel
Catalog No. NC9130814