Learn More
Medchemexpress LLC NS1643 | 448895-37-2 | 99.04% | 380.24 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16916
Encompass_Preferred
NS1643 is a partial agonist of human ether-a-go-go-related gene (hERG) K+ channels with an EC50 of 10.5 μM. It inhibits the growth of breast cancer tumors in TNBC mouse models and also inhibits cell migration and invasion of breast cancer cells.
  • Partial agonist of hERG K+ channels (EC50 of 10.5 μM)
  • Inhibits breast cancer tumor growth in TNBC mouse models
  • Inhibits cell migration and invasion of breast cancer cells
  • Dose- and voltage-dependently increases hERG current in oocytes
  • Slows the rate of hERG inactivation and reduces hERG channel rectification
  • Decreases Vimentin, N-cadherin, and CD44 levels, and increases E-cadherin in breast cancer cell lines (MDA-MB-231, SKBR3, and MCF7)
  • Inhibits breast tumor metastasis in NSG mice with human-derived TNBC tumor xenografts
Catalog No. 50-003-95300