Learn More
Medchemexpress LLC HY-110153 10mg Medchemexpress, NS19504 CAS:327062-46-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11015310MG
Encompass_Preferred
Medchemexpress, HY-110153 10mg NS19504 CAS:327062-46-4 NS19504 is a Ca2+-activated K+ channel (BK channel, KCa1.1 channel) activator (EC50=11.0 µM) with relaxing effect on bladder smooth muscle spontaneous phasic contractions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8755